MilliporeSigma
β₂-glycoprotein I: a novel component of innate immunity.
Blood (2011-04-02)
Cetin Agar, Philip G de Groot, Matthias Mörgelin, Stephanie D D C Monk, Gwendolyn van Os, Johannes H M Levels, Bas de Laat, Rolf T Urbanus, Heiko Herwald, Tom van der Poll, Joost C M Meijers
ABSTRACT
Sepsis is a systemic host response to invasive infection by bacteria. Despite treatment with antibiotics, current mortality rates are in the range of 20%-25%, which makes sepsis the most important cause of death in intensive care. Gram-negative bacteria are a prominent cause of sepsis. Lipopolysaccharide (LPS), one of the major constituents of the outer membrane of Gram-negative bacteria, plays a major role in activating the host's immune response by binding to monocytes and other cells. Several proteins are involved in neutralization and clearance of LPS from the bloodstream. Here, we provide evidence that β₂-glycoprotein I (β₂GPI) is a scavenger of LPS. In vitro, β₂GPI inhibited LPS-induced expression of tissue factor and IL-6 from monocytes and endothelial cells. Binding of β₂GPI to LPS caused a conformational change in β₂GPI that led to binding of the β₂GPI-LPS complex to monocytes and ultimately clearance of this complex. Furthermore, plasma levels of β₂GPI were inversely correlated with temperature rise and the response of inflammatory markers after a bolus injection of LPS in healthy individuals. Together, these observations provide evidence that β₂GPI is involved in the neutralization and clearance of LPS and identify β₂GPI as a component of innate immunity.
Product Description
Millipore
Ultrafree® Centrifugal Filter, 0.5 mL Sample Volume, pore size 0.22 μm, PVDF membrane (hydrophilic)