MilliporeSigma
  • Development and optimization of a useful assay for determining Hsp90's inherent ATPase activity.
Development and optimization of a useful assay for determining Hsp90's inherent ATPase activity.
Bioorganic & medicinal chemistry (2005-10-11)
Christopher Avila, Boris A Kornilayev, Brian S J Blagg
ABSTRACT
The Hsp90 molecular chaperone is responsible for the conformational maturation of nascent polypeptides and the rematuration of denatured proteins. Inhibition of Hsp90 represents a promising approach towards the treatment of cancer because numerous signaling cascades can be simultaneously targeted by disruption of the Hsp90-mediated process. Hsp90's ATPase activity is essential to the Hsp90-mediated protein folding process, consequently, a coupled assay was developed and optimized for determination of Hsp90's inherent ATPase activity. Using maltose phosphorylase, glucose oxidase, and horseradish peroxidase as components of this assay, a highly reproducible assay with a Z-factor of 0.87 has been produced.
Sigma-Aldrich
Maltose Phosphorylase from Enterococcus sp., recombinant, expressed in E. coli, lyophilized powder