MilliporeSigma
  • N-terminal processing: the methionine aminopeptidase and N alpha-acetyl transferase families.
N-terminal processing: the methionine aminopeptidase and N alpha-acetyl transferase families.
Trends in biochemical sciences (1998-08-11)
R A Bradshaw, W W Brickey, K W Walker
ABSTRACT
Removal of the initiator methionine and/or acetylation of the alpha-amino group are among the earliest possible chemical modifications that occur during protein synthesis in eukaryotes. These events are catalyzed by methionine aminopeptidase and N alpha-acetyltransferase, respectively. Recent advances in the isolation and characterization of these enzymes indicate that they exist as isoforms that vary in cellular location, function, and evolutionary origins.
Sigma-Aldrich
Methionine Aminopeptidase from Pyrococcus furiosus, ≥93% (SDS-PAGE), recombinant, expressed in E. coli