MilliporeSigma
  • c-Maf and JunB mediation of Th2 differentiation induced by the type 2 G protein-coupled receptor (VPAC2) for vasoactive intestinal peptide.
c-Maf and JunB mediation of Th2 differentiation induced by the type 2 G protein-coupled receptor (VPAC2) for vasoactive intestinal peptide.
Journal of immunology (Baltimore, Md. : 1950) (2004-06-10)
Julia Voice, Samantha Donnelly, Glenn Dorsam, Gregory Dolganov, Sudhir Paul, Edward J Goetzl
ABSTRACT
Vasoactive intestinal peptide and its G protein-coupled receptors, VPAC(1) and VPAC(2), regulate critical aspects of innate and adaptive immunity. T cell VPAC(2)Rs mediate changes in cytokine generation, which potently increase the Th2/Th1 ratio and consequently shift the effector responses toward allergy and inflammation. To examine mechanisms of VPAC(2) promotion of the Th2 phenotype, we analyzed controls of IL-4 transcription in CD4 T cells from T cell-targeted VPAC(2) transgenic (Tg), VPAC(2) knockout, and wild-type (WT) mice. c-maf and junB mRNA, protein, and activity were significantly up-regulated to a higher level in TCR-stimulated CD4 T cells from Tg mice compared with those from knockout and WT C57BL/6 mice. In contrast, GATA3, T-bet, and NFATc levels were identical in WT and Tg CD4 T cells. Vasoactive intestinal peptide binding to VPAC(2) on CD4 T cells specifically induces an up-regulation of the Th2-type transcription factors c-Maf and JunB, which consequently enhances IL-4 and IL-5 production, leading to a Th2-type phenotype.
Sigma-Aldrich
ChemiSCREEN Human VPAC2 VIP Receptor Membrane Preparation, Human VPAC2 / VIP2 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.