MilliporeSigma
  • Isolation and characterization of a homogeneous isoenzyme of wheat germ acid phosphatase.
Isolation and characterization of a homogeneous isoenzyme of wheat germ acid phosphatase.
Archives of biochemistry and biophysics (1991-08-01)
P P Waymack, R L Van Etten
ABSTRACT
An acid phosphatase (orthophosphoric monoester phosphohydrolase, acid optimum; EC 3.1.3.2) isoenzyme from wheat germ was purified 7000-fold to homogeneity. The effect of wheat germ sources and their relationship to the isoenzyme content and purification behavior of acid phosphatases was investigated. Extensive information about the purification and stabilization of the enzyme is provided. The instability of isoenzymes in the latter stages of purification appeared to be the result of surface inactivation together with a sensitivity to dilution that could be partially offset by addition of Triton X-100 during chromatographic procedures. Added sulfhydryl protecting reagents had no effect on activity or stability, which was greatest in the pH range 4-7. The purified isoenzyme was homogeneous by polyacrylamide gel electrophoresis and exhibited the highest specific activity and turnover number reported for any acid phosphatase. The molecular weights of the pure isoenzyme and of related isoenzymes from wheat germ were found to be identical (58,000). The pure isoenzyme contained a single polypeptide chain and had a negligible carbohydrate content. The amino acid composition was determined. Of the various reasons that were considered to explain isoenzyme occurrence, a genetic basis was considered most likely. The enzyme was found to exhibit substrate inhibition with some substrates below pH 6, while above pH 8 it exhibited downwardly curving Lineweaver-Burk plots of the type that are generally described as "substrate activation". The observation of a phosphotransferase activity was consistent with the formation of a covalent phosphoenzyme intermediate, while inactivation by diethyl pyrocarbonate was consistent with the presence of an active site histidine.
Sigma-Aldrich
Phosphatase, Acid from potato, lyophilized powder, ≥3.0 units/mg solid
Pricing and availability is not currently available.