MilliporeSigma
Avoiding false positives with PCR.
Nature (1989-05-18)
S Kwok, R Higuchi
ABSTRACT
The exquisite sensitivity of the polymerase chain reaction means DNA contamination can ruin an entire experiment. Tidiness and adherence to a strict set of protocols can avoid disaster.
Sigma-Aldrich
JumpStart Taq ReadyMix, Complete optimized reagent for hot-start PCR at 2X concentration
Pricing and availability is not currently available.