MilliporeSigma
  • Laminin alpha2 chain-positive vessels and epidermal growth factor in lung neuroendocrine carcinoma: a model of a novel cooperative role of laminin-2 and epidermal growth factor in vessel neoplastic invasion and metastasis.
Laminin alpha2 chain-positive vessels and epidermal growth factor in lung neuroendocrine carcinoma: a model of a novel cooperative role of laminin-2 and epidermal growth factor in vessel neoplastic invasion and metastasis.
The American journal of pathology (2006-03-02)
Domenico Vitolo, Luciano Ciocci, Gloria Deriu, Silvia Spinelli, Stefania Cortese, Laura Masuelli, Stefania Morrone, Mary Jo Filice, Giorgio Furio Coloni, Pier Giorgio Natali, Carlo Davide Baroni
ABSTRACT
Capillaries expressing the laminin alpha2 chain in basement membranes may be considered early developing vessels in normal and neoplastic human tissues. Therefore, we investigated whether up-regulation of this extracellular matrix protein favors transendothelial migration of neoplastic cells and then metastasis. In lung small and large cell neuroendocrine carcinomas, which exhibit a stronger metastatic tendency among carcinomas, laminin alpha2 chain-positive vessels were more numerous than in carcinoid tumors and supraglottis, breast, and lung non-small cell carcinomas, suggesting a direct relationship between these vessels and metastasis. In vitro studies showed that epidermal growth factor (EGF) induced a more efficient migration of the AE-2 lung neuroendocrine carcinoma cell line through the purified laminin alpha2 chain rather than through the laminin beta1 chain and fibronectin. AE-2 cells constitutively expressed all EGF receptors and the alpha6beta1 integrin, which is one of the laminin alpha2 chain receptors. EGF up-regulated alpha6beta1 expression in several tumors. In this regard, we show that EGF increased the chemo-kinetic migration of AE-2 cells through EAHY endothelial monolayers, which was inhibited by the anti-alpha6 integrin chain monoclonal antibody. These data indicate that laminin alpha2 chain and alpha6beta1 may be mutually involved in EGF-dependent migration of AE-2 cells and that laminin alpha2 chain-positive vessels may favor metastasis of EGF-dependent tumors.
Sigma-Aldrich
Human Laminin (pepsinized) Purified Protein
Pricing and availability is not currently available.