MilliporeSigma
  • Arachidonic acid stimulates extracellular Ca(2+) entry in rat pancreatic beta cells via activation of the noncapacitative arachidonate-regulated Ca(2+) (ARC) channels.
Arachidonic acid stimulates extracellular Ca(2+) entry in rat pancreatic beta cells via activation of the noncapacitative arachidonate-regulated Ca(2+) (ARC) channels.
Cell calcium (2009-12-19)
Valerie Yeung-Yam-Wah, Andy K Lee, Frederick W Tse, Amy Tse
ABSTRACT
Arachidonic acid (AA) is generated in the pancreatic islets during glucose stimulation. We investigated whether AA activated extracellular Ca(2+) entry in rat pancreatic beta cells via a pathway that was independent of the activation of voltage-gated Ca(2+) channels. The AA triggered [Ca(2+)](i) rise did not involve activation of GPR40 receptors or AA metabolism. When cells were voltage clamped at -70mV, the AA-mediated intracellular Ca(2+) release was accompanied by extracellular Ca(2+) entry. AA accelerated the rate of Mn(2+) quench of indo-1 fluorescence (near the Ca(2+)-independent wavelength of indo-1), reflecting the activation of a Ca(2+)-permeable pathway. The AA-mediated acceleration of Mn(2+) quench was inhibited by La(3+) but not by 2-APB (a blocker of capacitative Ca(2+) entry), suggesting the involvement of arachidonate-regulated Ca(2+) (ARC) channels. Consistent with this, intracellular application of the charged membrane-impermeant analog of AA, arachidonyl-coenzyme A (ACoA) triggered extracellular Ca(2+) entry, as well as the activation of a La(3+)-sensitive small inward current (1.7pA/pF) at -70mV. Our results indicate that the activation of ARC channels by intracellular AA triggers extracellular Ca(2+) entry. This action may contribute to the effects of AA on Ca(2+) signals and insulin secretion in rat beta cells.
Sigma-Aldrich
Arachidonoyl coenzyme A lithium salt, ≥85%
Pricing and availability is not currently available.