MilliporeSigma
  • Isolation and sequence determination of an active site peptide of rabbit muscle pyruvate kinase.
Isolation and sequence determination of an active site peptide of rabbit muscle pyruvate kinase.
Archives of biochemistry and biophysics (1987-02-15)
G Bezares, J Eyzaguirre, M V Hinrichs, R L Heinrikson, I Reardon, R G Kemp, S P Latshaw, S Bazaes
ABSTRACT
Rabbit muscle pyruvate kinase was inactivated by 2', 3'-dialdehyde ADP with the incorporation of one molecule of reagent per enzyme subunit. The inactivated protein was digested with trypsin after reduction and carboxymethylation. The labeled peptide was isolated by gel filtration and further purified by HPLC. The peptide was sequenced both by liquid-phase and gas-phase automatic Edman degradation. A 34-residue peptide was obtained. This peptide is identical to a tryptic peptide labeled with trinitrobenzenesulfonate, isolated and sequenced by Johnson et al. (Biochem. Biophys. Res. Commun. (1979) 90, 525-530) from bovine muscle pyruvate kinase. Available evidence suggests that dialdehyde ADP labels the enzyme at the same lysine in position 25 of the peptide, as found by Johnson et al. The high homology between the isolated peptide and regions of other pyruvate kinases from low to high eukaryotes supports the idea that this peptide is related to the enzyme active site.
Sigma-Aldrich
Adenosine 5′-diphosphate, periodate oxidized sodium salt, 90-95%