MilliporeSigma
  • Effects of astaxanthin on lipopolysaccharide-induced inflammation in vitro and in vivo.
Effects of astaxanthin on lipopolysaccharide-induced inflammation in vitro and in vivo.
Investigative ophthalmology & visual science (2003-05-27)
Kazuhiro Ohgami, Kenji Shiratori, Satoshi Kotake, Tomomi Nishida, Nobuhisa Mizuki, Kazunaga Yazawa, Shigeaki Ohno
ABSTRACT
Astaxanthin (AST) is a carotenoid that is found in marine animals and vegetables. Several previous studies have demonstrated that AST exhibits a wide variety of biological activities including antioxidant, antitumor, and anti-Helicobacter pylori effects. In this study, attention was focused on the antioxidant effect of AST. The object of the present study was to investigate the efficacy of AST in endotoxin-induced uveitis (EIU) in rats. In addition, the effect of AST on endotoxin-induced nitric oxide (NO), prostaglandin E2 (PGE2), and tumor necrosis factor (TNF)-alpha production in a mouse macrophage cell line (RAW 264.7) was studied in vitro. EIU was induced in male Lewis rats by a footpad injection of lipopolysaccharide (LPS). AST or prednisolone was administered intravenously at 30 minutes before, at the same time as, or at 30 minutes after LPS treatment. The number of infiltrating cells and protein concentration in the aqueous humor collected at 24 hours after LPS treatment was determined. RAW 264.7 cells were pretreated with various concentrations of AST for 24 hours and subsequently stimulated with 10 microg/mL of LPS for 24 hours. The levels of PGE2, TNF-alpha, and NO production were determined in vivo and in vitro. AST suppressed the development of EIU in a dose-dependent fashion. The anti-inflammatory effect of 100 mg/kg AST was as strong as that of 10 mg/kg prednisolone. AST also decreased production of NO, activity of inducible nitric oxide synthase (NOS), and production of PGE2 and TNF-alpha in RAW264.7 cells in vitro in a dose-dependent manner. This study suggests that AST has a dose-dependent ocular anti-inflammatory effect, by the suppression of NO, PGE2, and TNF-alpha production, through directly blocking NOS enzyme activity.
Sigma-Aldrich
Griess reagent (modified)
Pricing and availability is not currently available.