MilliporeSigma
  • Restoration of norepinephrine and reversal of phenotypes in mice lacking dopamine beta-hydroxylase.
Restoration of norepinephrine and reversal of phenotypes in mice lacking dopamine beta-hydroxylase.
Journal of neurochemistry (1998-05-29)
S A Thomas, B T Marck, R D Palmiter, A M Matsumoto
ABSTRACT
Mice with a targeted disruption of the dopamine beta-hydroxylase (DBH) gene are unable to synthesize norepinephrine (NE) and epinephrine. These mice have elevated levels of dopamine in most tissues, although the levels are only a fraction of those normally found for NE. It is noteworthy that NE can be restored to normal levels in many tissues after a single injection of the synthetic amino acid precursor of NE, L-threo-3,4-dihydroxyphenylserine (DOPS). In other tissues, NE can be restored to normal levels after multiple injections of DOPS, whereas in the midbrain and cerebellum, restoration of NE is limited to 25-30% of normal. NE levels typically peak approximately 5 h after DOPS administration and are undetectable by 48 h. Epinephrine levels are more difficult to restore. The elevated levels of dopamine fall modestly after injection of DOPS. S(-)-Carbidopa, which does not cross the blood-brain barrier, inhibits aromatic L-amino acid decarboxylase and effectively prevents restoration of NE by DOPS in the periphery, while allowing restoration in the CNS. Ptosis and reductions in male fertility, hind-limb extension, postdecapitation convulsions, and uncoupling protein expression in dopamine beta-hydroxylase-deficient mice are all reversed by DOPS injection.
Sigma-Aldrich
DL-3,4-Dihydroxyphenylserine, ≥98% (HPLC), powder
Pricing and availability is not currently available.