MilliporeSigma
  • Enhancing the thermal stability of polythiophene:fullerene solar cells by decreasing effective polymer regioregularity.
Enhancing the thermal stability of polythiophene:fullerene solar cells by decreasing effective polymer regioregularity.
Journal of the American Chemical Society (2006-10-26)
Kevin Sivula, Christine K Luscombe, Barry C Thompson, Jean M J Fréchet
ABSTRACT
Regioregularity has been shown to be an important aspect for the properties of poly(3-hexylthiophene) both in the pristine material and in bulk heterojunction photovoltaics with the fullerene derivative PCBM. Here we present a straightforward method to adjust the effective regioregularity of poly(3-hexylthiophene) by introducing 3,4-dihexylthiophene into the polymer chain. By investigating bulk heterojunction morphology and photovoltaic performance upon annealing, we observe that polythiophene with 91% regioregularity maintains equivalent electronic properties but forms a more thermally stable interpenetrating network with PCBM than polythiophene with regioregularity >96%.
Sigma-Aldrich
3,4-Dihexylthiophene, 97%