MilliporeSigma
  • Enantioselective bromoaminocyclization using amino-thiocarbamate catalysts.
Enantioselective bromoaminocyclization using amino-thiocarbamate catalysts.
Journal of the American Chemical Society (2011-05-05)
Ling Zhou, Jie Chen, Chong Kiat Tan, Ying-Yeung Yeung
ABSTRACT
A facile and efficient enantioselective bromoaminocyclization of unsaturated sulfonamides has been developed using an amino-thiocarbamate catalyst. A range of enantioenriched pyrrolidines were prepared with up to 99% yield and 99% ee. The corresponding lactams could be obtained through oxidation of the pyrrolidines.
Sigma-Aldrich
Methylboronic acid, 97%