MilliporeSigma
  • Estrogen-dependent gene regulation by an oxidative metabolite of diethylstilbestrol, diethylstilbestrol-4',4"-quinone.
Estrogen-dependent gene regulation by an oxidative metabolite of diethylstilbestrol, diethylstilbestrol-4',4"-quinone.
Steroids (1998-04-29)
K Chae, J Lindzey, J A McLachlan, K S Korach
ABSTRACT
Diethylstilbestrol (DES) is a well-characterized carcinogen in humans and animals although its mechanisms of carcinogenicity are not yet known. While the estrogenic activity of DES is important, there is evidence that oxidative metabolism also plays an important role for its toxicity. DES is oxidatively metabolized in vivo and in vitro to a number of compounds including diethylstilbestrol-4',4"-quinone (DQ), an unstable and reactive intermediate, and Z,Z-dienestrol (ZZ-DIEN). Estrogen receptor (ER) binding assays with mouse uterine cytosol indicate that DES, DQ and ZZ-DIEN have relative binding affinities of 286, 3.6 and 0.3, respectively, relative to estradiol as 100. In addition, DQ binds irreversibly and specifically to ER suggesting that DQ may be biologically active despite its rapid metabolism and lower binding affinity compared to DES. To test this, COS-1 cells were transfected with an estrogen responsive reporter construct containing of VitA2 estrogen response element (ERE) with or without an ER expression vector. In the presence of ER, treatments with DES, DQ and ZZ-DIEN resulted in 11, 10, and 2-fold induction of chloramphenicol acetyltransferase (CAT) activity, respectively. This induction was mediated by estrogen receptor since it was suppressed by pretreatment with a 10-fold excess of the pure antiestrogen ICI 182,780. These data indicate that DQ is a biologically active intermediate that is capable of transactivation of estrogen responsive genes through the ER. Furthermore, the data suggest that the ability of DQ to irreversibly bind ER may result in persistent stimulation of ER. This persistent stimulation may be related to the carcinogenicity of DES.