MilliporeSigma
  • P1,P4-diadenosine 5' tetraphosphate induces nitric oxide release from bovine aortic endothelial cells.
P1,P4-diadenosine 5' tetraphosphate induces nitric oxide release from bovine aortic endothelial cells.
FEBS letters (1998-06-24)
R H Hilderman, E F Christensen
ABSTRACT
Since the infusion of P1,P4-diadenosine 5' tetraphosphate (Ap4A) into animal models induces vasodilation [1,2], the present study was performed to determine whether Ap4A induces the release of nitric oxide (NO) from endothelial cells. Ap4A induced NO release was 4.2-fold greater than the amount of NO released under basal condition. Ap4A induced NO release was inhibited by NG-nitro-L-arginine (L-NNA) and this inhibition was reversed by L-Arg. In addition, EGTA inhibits Ap4A induced NO release. These data are consistent with Ap4A inducing the release of NO from endothelial cells through the activation of endothelial nitric oxide synthase.
Sigma-Aldrich
P1,P4-Di(adenosine-5′) tetraphosphate ammonium salt, ≥95% (HPLC), powder