MilliporeSigma
Adenine dinucleotides: a novel class of signalling molecules.
Journal of autonomic pharmacology (1996-12-01)
A Ogilvie, R Bläsius, E Schulze-Lohoff, R B Sterzel
ABSTRACT
1. Adenine dinucleotides (Ap3A, Ap4A, Ap5A, Ap6A) are stored in secretory granules of thrombocytes, chromaffin cells and neuronal cells. After release into the extracellular space, the dinucleotides exhibit divergent biological effects on a variety of target cells and organs. The dinucleotides are metabolized by soluble enzymes in the blood plasma as well as by membrane-bound ectoenzymes of endothelial cells, smooth muscle cells, and other cell types. 2. The enzymatic cleavage of the dinucleotides plays a dual role for their biological function: (a) termination of the signal; and (b) generation of purinergically active products such as ATP, ADP and finally adenosine. In contrast to ATP the dinucleotides are long-lived purine nucleotides in the blood. 3. The potential role of the dinucleotides as signalling molecules has been demonstrated in several systems. The adenosine polyphosphates have autocrine function for thrombocytes. Ap3A at low concentration reversibly activates isolated platelets. The mechanism of activation has been elucidated by showing a continuous cleavage of Ap3A, leading to the formation of ADP which is a known agonist of the P2T receptor on thrombocytes. Ap4A and other dinucleotides act as antagonists and inhibit platelet activation. 4. The vasotone of perfused isolated arteries as well as of resistance vessels in the beating heart is differentially influenced by adenine dinucleotides. While Ap3A and Ap4A exhibit relaxing effects at micromolar concentrations, Ap5A and Ap6A elicit vasoconstriction in these vessels. 5. In rat kidney mesangial cells adenine dinucleotides efficiently promote growth. Stimulation of DNA synthesis by various growth factors is enhanced synergistically. ApnA significantly increase the expression of the early growth response gene Egr-1. 6. The specificity and, in some tissues, the uniqueness of effects evoked by dinucleotides may be mediated by genuine dinucleotide receptors (P4) or by specialized P2 receptors (P2D).
Sigma-Aldrich
P1,P4-Di(adenosine-5′) tetraphosphate ammonium salt, ≥95% (HPLC), powder