MilliporeSigma
  • Measurement and correlation of solubility of meropenem trihydrate in binary (water+ acetone/tetrahydrofuran) solvent mixtures
Measurement and correlation of solubility of meropenem trihydrate in binary (water+ acetone/tetrahydrofuran) solvent mixtures
Chinese Journal of Chemical Engineering (2017)
Zhou L, et al.