MilliporeSigma
  • Synthesis of an allyl carbonate monomer as alternative to TEGDMA in the formulation of dental composite resins.
Synthesis of an allyl carbonate monomer as alternative to TEGDMA in the formulation of dental composite resins.
Journal of the mechanical behavior of biomedical materials (2018-08-03)
Carlos E Cuevas-Suárez, J Abraham González-López, Adriana Fernandes da Silva, Evandro Piva, Ana M Herrera-González
ABSTRACT
Over the past years, significant effort has been dedicated to synthesizing low-shrinking formulations, however, development of dental composites with low volumetric shrinkage continues to be challenging. The purpose of this study was to synthesize a bisphenol allylic derivate (BPhADAC) and evaluate its inclusion in the formulation of a photopolymerizable dental composite resin, as a BisGMA diluent. Experimental (BisGMA/BPhADAC) and control (BisGMA/TEGDMA) photopolymerizable composites were prepared. Double bond conversion, polymerization kinetics, volumetric shrinkage, water sorption, solubility, and flexural properties were investigated. The experimental composite showed higher degree of conversion values, less volumetric shrinkage and less water sorption than the control composite (p < 0.05). In addition, flexural strength between the materials was found to be similar. The overall properties prove that the allylic monomer BPhADAC could be potentially useful in the formulation of low-shrinking dental composite resins.
Sigma-Aldrich
Allyl chloroformate, 97%
Pricing and availability is not currently available.