MilliporeSigma
  • Gold(I)-catalyzed isomerization of allenyl carbinol esters: an efficient access to functionalized 1,3-butadien-2-ol esters.
Gold(I)-catalyzed isomerization of allenyl carbinol esters: an efficient access to functionalized 1,3-butadien-2-ol esters.
Organic letters (2007-02-14)
Andrea K Buzas, Florin M Istrate, Fabien Gagosz
ABSTRACT
A study concerning the gold(I)-catalyzed rearrangement of diversely substituted allenyl carbinol esters into functionalized 1,3-butadien-2-ol esters is described. The mild conditions employed allow the efficient, rapid, and stereoselective synthesis of a variety of such compounds via a new 1,3-shift of an ester moiety onto a gold-activated allene. [structure: see text]
Sigma-Aldrich
2-Dicyclohexylphosphino-2′,4′,6′-triisopropylbiphenyl gold(I) bis(trifluoromethanesulfonyl)imide
Pricing and availability is not currently available.