MilliporeSigma
  • Renoprotective effect of Zhenwu decoction against renal fibrosis by regulation of oxidative damage and energy metabolism disorder.
Renoprotective effect of Zhenwu decoction against renal fibrosis by regulation of oxidative damage and energy metabolism disorder.
Scientific reports (2018-10-04)
Shasha Li, Xue Xiao, Ling Han, Yiming Wang, Guoan Luo
ABSTRACT
Zhenwu decoction (ZWD) is a promising traditional Chinese prescription against renal fibrosis, while its underlying mechanism remains unclear. Rat model of renal fibrosis were established and divided into control group, model group, ZWD treatment group and enalapril maleate treatment group. Metabolic profiles on serum samples from each group were acquired by using ultra performance liquid chromatography coupled with quadrupole time-of-flight high-resolution mass spectrometry. Metabolomics combined with molecular biology were comparatively conducted on samples of various groups. Fifteen potential biomarkers were identified and these biomarkers are mainly phospholipids and fatty acids. The results showed renal fibrosis was associated with oxidative damage and energy metabolism disorder. The results of histopathology, biochemistry and metabolomics demonstrated that ZWD exhibited an efficient renoprotective effect by alleviating oxidative stress, increasing energy metabolism and regulating fibrotic cytokines. This study provided scientific support for the research and development of new drugs from traditional Chinese medicine.
Sigma-Aldrich
9(S)-HODE, ≥98% (HPLC)