MilliporeSigma
  • Assembly of Tissue-Engineered Blood Vessels with Spatially Controlled Heterogeneities.
Assembly of Tissue-Engineered Blood Vessels with Spatially Controlled Heterogeneities.
Tissue engineering. Part A (2018-05-05)
Hannah A Strobel, Tracy A Hookway, Marco Piola, Gianfranco Beniamino Fiore, Monica Soncini, Eben Alsberg, Marsha W Rolle
ABSTRACT
Tissue-engineered human blood vessels may enable in vitro disease modeling and drug screening to accelerate advances in vascular medicine. Existing methods for tissue-engineered blood vessel (TEBV) fabrication create homogenous tubes not conducive to modeling the focal pathologies characteristic of certain vascular diseases. We developed a system for generating self-assembled human smooth muscle cell (SMC) ring units, which were fused together into TEBVs. The goal of this study was to assess the feasibility of modular assembly and fusion of ring building units to fabricate spatially controlled, heterogeneous tissue tubes. We first aimed to enhance fusion and reduce total culture time, and determined that reducing ring preculture duration improved tube fusion. Next, we incorporated electrospun polymer ring units onto tube ends as reinforced extensions, which allowed us to cannulate tubes after only 7 days of fusion, and culture tubes with luminal flow in a custom bioreactor. To create focal heterogeneities, we incorporated gelatin microspheres into select ring units during self-assembly, and fused these rings between ring units without microspheres. Cells within rings maintained their spatial position along tissue tubes after fusion. Because tubes fabricated from primary SMCs did not express contractile proteins, we also fabricated tubes from human mesenchymal stem cells, which expressed smooth muscle alpha actin and SM22-α. This work describes a platform approach for creating modular TEBVs with spatially defined structural heterogeneities, which may ultimately be applied to mimic focal diseases such as intimal hyperplasia or aneurysm.
Sigma-Aldrich
2,2,2-Trifluoroethanol, ReagentPlus®, ≥99%
Pricing and availability is not currently available.