MilliporeSigma
  • Induction of Neoantigen-Specific Cytotoxic T Cells and Construction of T-cell Receptor-Engineered T Cells for Ovarian Cancer.
Induction of Neoantigen-Specific Cytotoxic T Cells and Construction of T-cell Receptor-Engineered T Cells for Ovarian Cancer.
Clinical cancer research : an official journal of the American Association for Cancer Research (2018-05-04)
Tatsuo Matsuda, Matthias Leisegang, Jae-Hyun Park, Lili Ren, Taigo Kato, Yuji Ikeda, Makiko Harada, Kazuma Kiyotani, Ernst Lengyel, Gini F Fleming, Yusuke Nakamura
ABSTRACT
Purpose: Current evolution of cancer immunotherapies, such as immune checkpoint blockade, has implicated neoantigens as major targets of anticancer cytotoxic T cells. Adoptive T-cell therapy with neoantigen-specific T-cell receptor (TCR)-engineered T cells would be an attractive therapeutic option for advanced cancers where the host antitumor immune function is strongly inhibited. We previously developed a rapid and efficient pipeline for production of neoantigen-specific TCR-engineered T cells using peripheral blood from an HLA-matched healthy donor. Our protocol required only 2 weeks from stimulation of T cells with neoantigen-loaded dendritic cells to the identification of neoantigen-specific TCRs. We conducted the pilot study to validate our protocol.Experimental Design: We used tumors from 7 ovarian cancer patients to validate our protocol.Results: We chose 14 candidate neoantigens from 7 ovarian tumors (1-3 candidates for each patient) and then successfully induced three neoantigen-specific T cells from 1 healthy donor and identified their TCR sequences. Moreover, we validated functional activity of the three identified TCRs by generating TCR-engineered T cells that recognized the corresponding neoantigens and showed cytotoxic activity in an antigen dose-dependent manner. However, one case of neoantigen-specific TCR-engineered T cells showed cross-reactivity against the corresponding wild-type peptide.Conclusions: This pilot study demonstrated the feasibility of our efficient process from identification of neoantigen to production of the neoantigen-targeting cytotoxic TCR-engineered T cells for ovarian cancer and revealed the importance of careful validation of neoantigen-specific TCR-engineered T cells to avoid severe immune-related adverse events. Clin Cancer Res; 24(21); 5357-67. ©2018 AACRSee related commentary by Anczurowski and Hirano, p. 5195.
Sigma-Aldrich
Bovine IL2 / Interleukin-2 ELISA Kit, for serum, plasma and cell culture supernatants