MilliporeSigma
  • Dynamic coordination of two-metal-ions orchestrates λ-exonuclease catalysis.
Dynamic coordination of two-metal-ions orchestrates λ-exonuclease catalysis.
Nature communications (2018-10-26)
Wonseok Hwang, Jungmin Yoo, Yuno Lee, Suyeon Park, Phuong Lien Hoang, HyeokJin Cho, Jeongmin Yu, Thi Minh Hoa Vo, Minsang Shin, Mi Sun Jin, Daeho Park, Changbong Hyeon, Gwangrog Lee
ABSTRACT
Metal ions at the active site of an enzyme act as cofactors, and their dynamic fluctuations can potentially influence enzyme activity. Here, we use λ-exonuclease as a model enzyme with two Mg2+ binding sites and probe activity at various concentrations of magnesium by single-molecule-FRET. We find that while MgA2+ and MgB2+ have similar binding constants, the dissociation rate of MgA2+ is two order of magnitude lower than that of MgB2+ due to a kinetic-barrier-difference. At physiological Mg2+ concentration, the MgB2+ ion near the 5'-terminal side of the scissile phosphate dissociates each-round of degradation, facilitating a series of DNA cleavages via fast product-release concomitant with enzyme-translocation. At a low magnesium concentration, occasional dissociation and slow re-coordination of MgA2+ result in pauses during processive degradation. Our study highlights the importance of metal-ion-coordination dynamics in correlation with the enzymatic reaction-steps, and offers insights into the origin of dynamic heterogeneity in enzymatic catalysis.
Sigma-Aldrich
Lambda Phage DNA, Non-methylated from Escherichia coli host strain GM119 (rm-,dam-,dcm-), solution
Pricing and availability is not currently available.