MilliporeSigma
  • Identification and antitumor activity of a novel inhibitor of the NIMA-related kinase NEK6.
Identification and antitumor activity of a novel inhibitor of the NIMA-related kinase NEK6.
Scientific reports (2018-10-31)
Marta De Donato, Benedetta Righino, Flavia Filippetti, Alessandra Battaglia, Marco Petrillo, Davide Pirolli, Giovanni Scambia, Maria Cristina De Rosa, Daniela Gallo
ABSTRACT
The NIMA (never in mitosis, gene A)-related kinase-6 (NEK6), which is implicated in cell cycle control and plays significant roles in tumorigenesis, is an attractive target for the development of novel anti-cancer drugs. Here we describe the discovery of a potent ATP site-directed inhibitor of NEK6 identified by virtual screening, adopting both structure- and ligand-based techniques. Using a homology-built model of NEK6 as well as the pharmacophoric features of known NEK6 inhibitors we identified novel binding scaffolds. Twenty-five compounds from the top ranking hits were subjected to in vitro kinase assays. The best compound, i.e. compound 8 ((5Z)-2-hydroxy-4-methyl-6-oxo-5-[(5-phenylfuran-2-yl)methylidene]-5,6-dihydropyridine-3-carbonitrile), was able to inhibit NEK6 with low micromolar IC50 value, also displaying antiproliferative activity against a panel of human cancer cell lines. Our results suggest that the identified inhibitor can be used as lead candidate for the development of novel anti-cancer agents, thus opening the possibility of new therapeutic strategies.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
0.2 mL
1 Available to ship on January 06, 2025
Details...
A recombinant, preservative-free antibody is available for your target. Try ZRB1312
¥98,300
100 μL
1 Available to ship on January 06, 2025
Details...
A recombinant, preservative-free antibody is available for your target. Try ZRB1312
¥98,300
0.5 mL
1 Available to ship on January 06, 2025
Details...
A recombinant, preservative-free antibody is available for your target. Try ZRB1312
¥195,000
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, Grade I, ≥99%, from microbial
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
0.2 mL
1 Available to ship on January 06, 2025
Details...
A recombinant, preservative-free antibody is available for your target. Try ZRB1312
¥98,300
100 μL
1 Available to ship on January 06, 2025
Details...
A recombinant, preservative-free antibody is available for your target. Try ZRB1312
¥98,300
0.5 mL
1 Available to ship on January 06, 2025
Details...
A recombinant, preservative-free antibody is available for your target. Try ZRB1312
¥195,000