MilliporeSigma
  • PPARβ Is Essential for Maintaining Normal Levels of PGC-1α and Mitochondria and for the Increase in Muscle Mitochondria Induced by Exercise.
PPARβ Is Essential for Maintaining Normal Levels of PGC-1α and Mitochondria and for the Increase in Muscle Mitochondria Induced by Exercise.
Cell metabolism (2017-05-04)
Jin-Ho Koh, Chad R Hancock, Shin Terada, Kazuhiko Higashida, John O Holloszy, Dong-Ho Han
ABSTRACT
The objective of this study was to evaluate the specific mechanism(s) by which PPARβ regulates mitochondrial content in skeletal muscle. We discovered that PPARβ increases PGC-1α by protecting it from degradation by binding to PGC-1α and limiting ubiquitination. PPARβ also induces an increase in nuclear respiratory factor 1 (NRF-1) expression, resulting in increases in mitochondrial respiratory chain proteins and MEF2A, for which NRF-1 is a transcription factor. There was also an increase in AMP kinase phosphorylation mediated by an NRF-1-induced increase in CAM kinase kinase-β (CaMKKβ). Knockdown of PPARβ resulted in large decreases in the levels of PGC-1α and mitochondrial proteins and a marked attenuation of the exercise-induced increase in mitochondrial biogenesis. In conclusion, PPARβ induces an increase in PGC-1α protein, and PPARβ is a transcription factor for NRF-1. Thus, PPARβ plays essential roles in the maintenance and adaptive increase in mitochondrial enzymes in skeletal muscle by exercise.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid