MilliporeSigma
  • Type I Interferon Inhibition of MicroRNA-146a Maturation Through Up-Regulation of Monocyte Chemotactic Protein-Induced Protein 1 in Systemic Lupus Erythematosus.
Type I Interferon Inhibition of MicroRNA-146a Maturation Through Up-Regulation of Monocyte Chemotactic Protein-Induced Protein 1 in Systemic Lupus Erythematosus.
Arthritis & rheumatology (Hoboken, N.J.) (2015-09-01)
Bo Qu, Jianchang Cao, Feifei Zhang, Huijuan Cui, Jialin Teng, Jia Li, Zheng Liu, Chris Morehouse, Bahija Jallal, Yuanjia Tang, Qiang Guo, Yihong Yao, Nan Shen
ABSTRACT
Systemic lupus erythematosus (SLE) is characterized by the uncontrolled production of inflammatory cytokines, among which type I interferon (IFN) is recognized as a crucial pathogenic factor. The expression of microRNA-146a (miR-146a) is reduced in the white blood cells of SLE patients and accounts for their overactivated inflammatory responses. However, the mechanism of the reduction of miR-146a is still not fully understood. This study was undertaken to test whether the key pathogenic cytokine, type I IFN, is responsible for the dysregulation of miR-146a in SLE. Gene and protein expression was measured in all cells by reverse transcription-quantitative polymerase chain reaction, Northern blotting, or Western blotting. In THP-1 cells, expression of monocyte chemotactic protein-induced protein 1 (MCPIP-1) was knocked down with a lentivirus encoding a short hairpin RNA targeting MCPIP1. The cells were pretreated with type I IFN and assessed for gene expression levels of miR-146a. White blood cells from patients with SLE were analyzed for the expression of the IFN-inducible genes MCPIP1 and miR-146a, and the gene expression data were compared for correlation. Pretreatment of THP-1 cells with type I IFN attenuated the induction of miR-146a posttranscriptionally, by down-regulating the expression of pre-miR-146a but not pri-miR-146a or its original unspliced transcript. Expression of MCPIP-1, which was enhanced by type I IFN, was found to be responsible for the inhibition of miR-146a. In white blood cells from patients with SLE, MCPIP1 expression was elevated, and its expression correlated positively with the IFN score and negatively with the miR-146a transcript level. Type I IFN inhibits the maturation of miR-146a through the up-regulation of MCPIP-1, and thus contributes to the uncontrolled inflammation and excessive inflammatory gene expression in SLE.
Sigma-Aldrich
Anti-ZC3H12A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution