MilliporeSigma
  • The chimeric TAC receptor co-opts the T cell receptor yielding robust anti-tumor activity without toxicity.
The chimeric TAC receptor co-opts the T cell receptor yielding robust anti-tumor activity without toxicity.
Nature communications (2018-08-05)
Christopher W Helsen, Joanne A Hammill, Vivian W C Lau, Kenneth A Mwawasi, Arya Afsahi, Ksenia Bezverbnaya, Lisa Newhook, Danielle L Hayes, Craig Aarts, Bojana Bojovic, Galina F Denisova, Jacek M Kwiecien, Ian Brain, Heather Derocher, Katy Milne, Brad H Nelson, Jonathan L Bramson
ABSTRACT
Engineering T cells with chimeric antigen receptors (CARs) is an effective method for directing T cells to attack tumors, but may cause adverse side effects such as the potentially lethal cytokine release syndrome. Here the authors show that the T cell antigen coupler (TAC), a chimeric receptor that co-opts the endogenous TCR, induces more efficient anti-tumor responses and reduced toxicity when compared with past-generation CARs. TAC-engineered T cells induce robust and antigen-specific cytokine production and cytotoxicity in vitro, and strong anti-tumor activity in a variety of xenograft models including solid and liquid tumors. In a solid tumor model, TAC-T cells outperform CD28-based CAR-T cells with increased anti-tumor efficacy, reduced toxicity, and faster tumor infiltration. Intratumoral TAC-T cells are enriched for Ki-67+ CD8+ T cells, demonstrating local expansion. These results indicate that TAC-T cells may have a superior therapeutic index relative to CAR-T cells.
Sigma-Aldrich
Monoclonal Anti-Cytokeratin, pan antibody produced in mouse, ascites fluid, clone PCK-26