MilliporeSigma
  • Novel histamine H3 receptor antagonists based on the 4-[(1H-imidazol-4-yl)methyl]piperidine scaffold.
Novel histamine H3 receptor antagonists based on the 4-[(1H-imidazol-4-yl)methyl]piperidine scaffold.
Bioorganic & medicinal chemistry letters (2005-10-26)
Wayne D Vaccaro, Rosy Sher, Michael Berlin, Neng-Yang Shih, Robert Aslanian, John H Schwerdt, Kevin D McCormick, John J Piwinski, Robert E West, John C Anthes, Shirley M Williams, Ren-Long Wu, H Susan She, Maria A Rivelli, Jennifer C Mutter, Michel R Corboz, John A Hey, Leonard Favreau
ABSTRACT
We report the discovery of novel histamine H(3) receptor antagonists based on 4-[(1H-imidazol-4-yl)methyl]piperidine. The most potent compounds in the series (e.g., 7) result from the attachment of a substituted aniline amide to the main pharmacophore piperidine via a two-methylene linker.
Sigma-Aldrich
3-Phenoxypropionic acid, 99%