MilliporeSigma
  • Pechini synthesis and characteristics of Gd2CoMnO6 nanostructures and its structural, optical and photocatalytic properties.
Pechini synthesis and characteristics of Gd2CoMnO6 nanostructures and its structural, optical and photocatalytic properties.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2018-06-25)
Reza Mohassel, Azam Sobhani, Masoud Salavati-Niasari, Mojgan Goudarzi
ABSTRACT
GCMO NSs were synthesized by the reaction of metal nitrate salts in the presence of stabilizing agent and PG, by a Pechini method. Citric acid, maleic acid, succinic acid and 1,3,5-benzenetricarboxylic acid were used as stabilizing agents. The structure, morphology, optical, magnetic and photocatalytic properties of the GCMO NSs were investigated using various characterization techniques. Effects of type of stabilizing agent, the molar ratio of stabilizing agent:PG and also calcination temperature on particle size and morphology of the products were investigated. Also the influence of kind of pollutant on photocatalytic behavior of GCMO NSs was evaluated.
Sigma-Aldrich
Manganese(II) nitrate hexahydrate, JIS special grade, ≥98.0%