MilliporeSigma
  • VCAM-1 upregulation accompanies muscle remodeling following resistance-type exercise in Snell dwarf (Pit1dw/dw ) mice.
VCAM-1 upregulation accompanies muscle remodeling following resistance-type exercise in Snell dwarf (Pit1dw/dw ) mice.
Aging cell (2018-07-12)
Erik P Rader, Marshall A Naimo, James Ensey, Brent A Baker
ABSTRACT
Snell dwarf mice (Pit1dw/dw ) exhibit deficiencies in growth hormone, prolactin, and thyroid stimulating hormone. Besides being an experimental model of hypopituitarism, these mice are long-lived (>40% lifespan extension) and utilized as a model of slowed/delayed aging. Whether this longevity is accompanied by a compromised quality of life in terms of muscular performance has not yet been characterized. In this study, we investigated nontrained and trained muscles 1 month following a general validated resistance-type exercise protocol in 3-month-old Snell dwarf mice and control littermates. Nontrained Snell dwarf gastrocnemius muscles exhibited a 1.3-fold greater muscle mass to body weight ratio than control values although muscle quality, maximum isometric torque normalized to muscle mass, and fatigue recovery were compromised. For control mice, training increased isometric torque (17%) without altering muscle mass. For Snell dwarf mice, isometric torque was unaltered by training despite decreased muscle mass that rendered muscle mass to body weight ratio comparable to control values. Muscle quality and fatigue recovery improved twofold and threefold, respectively, for Snell dwarf mice. This accompanied a fourfold increase in levels of vascular cell adhesion molecule-1 (VCAM-1), a mediator of progenitor cell recruitment, and muscle remodeling in the form of increased number of central nuclei, additional muscle fibers per unit area, and altered fiber type distribution. These results reveal a trade-off between muscle quality and longevity in the context of anterior pituitary hormone deficiency and that resistance-type training can diminish this trade-off by improving muscle quality concomitant with VCAM-1 upregulation and muscle remodeling.
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.