MilliporeSigma
A readily programmable, fully reversible shape-switching material.
Science advances (2018-08-29)
Matthew K McBride, Alina M Martinez, Lewis Cox, Marvin Alim, Kimberly Childress, Michael Beiswinger, Maciej Podgorski, Brady T Worrell, Jason Killgore, Christopher N Bowman
ABSTRACT
Liquid crystalline (LC) elastomers (LCEs) enable large-scale reversible shape changes in polymeric materials; however, they require intensive, irreversible programming approaches in order to facilitate controllable actuation. We have implemented photoinduced dynamic covalent chemistry (DCC) that chemically anneals the LCE toward an applied equilibrium only when and where the light-activated DCC is on. By using light as the stimulus that enables programming, the dynamic bond exchange is orthogonal to LC phase behavior, enabling the LCE to be annealed in any LC phase or in the isotropic phase with various manifestations of this capability explored here. In a photopolymerizable LCE network, we report the synthesis, characterization, and exploitation of readily shape-programmable DCC-functional LCEs to create predictable, complex, and fully reversible shape changes, thus enabling the literal square peg to fit into a round hole.
Sigma-Aldrich
3-Chloro-2-chloromethyl-1-propene, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium ethyl xanthogenate, 96%
Pricing and availability is not currently available.