MilliporeSigma
  • Targeting AURKA-CDC25C axis to induce synthetic lethality in ARID1A-deficient colorectal cancer cells.
Targeting AURKA-CDC25C axis to induce synthetic lethality in ARID1A-deficient colorectal cancer cells.
Nature communications (2018-08-12)
Changjie Wu, Junfang Lyu, Eun Ju Yang, Yifan Liu, Baoyuan Zhang, Joong Sup Shim
ABSTRACT
ARID1A, a component of the SWI/SNF chromatin remodeling complex, is a tumor suppressor with a high frequency of inactivating mutations in many cancers. Therefore, ARID1A deficiency has been exploited therapeutically for treating cancer. Here we show that ARID1A has a synthetic lethal interaction with aurora kinase A (AURKA) in colorectal cancer (CRC) cells. Pharmacological and genetic perturbations of AURKA selectively inhibit the growth of ARID1A-deficient CRC cells. Mechanistically, ARID1A occupies the AURKA gene promoter and negatively regulates its transcription. Cells lacking ARID1A show enhanced AURKA transcription, which leads to the persistent activation of CDC25C, a key protein for G2/M transition and mitotic entry. Inhibiting AURKA activity in ARID1A-deficient cells significantly increases G2/M arrest and induces cellular multinucleation and apoptosis. This study shows a novel synthetic lethality interaction between ARID1A and AURKA and indicates that pharmacologically inhibiting the AURKA-CDC25C axis represents a novel strategy for treating CRC with ARID1A loss-of-function mutations.
Sigma-Aldrich
MISSION® esiRNA, targeting human PLK1
Pricing and availability is not currently available.
Sigma-Aldrich
Imprint® Chromatin Immunoprecipitation Kit, Complete ChIP reaction in 6 hours in flexible strip well format
Pricing and availability is not currently available.
Sigma-Aldrich
Aurora-A Inhibitor I, ≥98% (HPLC)
Pricing and availability is not currently available.