MilliporeSigma
  • Tannic Acid/Fe3+ Nanoscaffold for Interfacial Polymerization: Toward Enhanced Nanofiltration Performance.
Tannic Acid/Fe3+ Nanoscaffold for Interfacial Polymerization: Toward Enhanced Nanofiltration Performance.
Environmental science & technology (2018-07-26)
Zhe Yang, Zhi-Wen Zhou, Hao Guo, Zhikan Yao, Xiao-Hua Ma, Xiaoxiao Song, Shien-Ping Feng, Chuyang Y Tang
ABSTRACT
Conventional thin-film composite (TFC) membranes suffer from the trade-off relationship between permeability and selectivity, known as the "upper bound". In this work, we report a high performance thin-film composite membrane prepared on a tannic acid (TA)-Fe nanoscaffold (TFCn) to overcome such upper bound. Specifically, a TA-Fe nanoscaffold was first coated onto a polysulfone substrate, followed by performing an interfacial polymerization reaction between trimesoyl chloride (TMC) and piperazine (PIP). The TA-Fe nanoscaffold enhanced the uptake of amine monomers and provided a platform for their controlled release. The smaller surface pore size of the TA-Fe coated substrate further eliminated the intrusion of polyamide into the substrate pores. The resulting membrane TFCn showed a water permeability of 19.6 ± 0.5 L m2- h-1 bar-1, which was an order of magnitude higher than that of control TFC membrane (2.2 ± 0.3 L m-2 h-1 bar-1). The formation of a more order polyamide rejection layer also significantly enhanced salt rejection (e.g., NaCl, MgCl2, Na2SO4, and MgSO4) and divalent to monovalent ion selectivity (e.g., NaCl/MgSO4). Compared to conventional TFC nanofiltration membranes, the novel TFCn membrane successfully overcame the longstanding permeability and selectivity trade-off. The current work paves a new avenue for fabricating high performance TFC membranes.
Sigma-Aldrich
1,3,5-Benzenetricarbonyl trichloride, 98%
Pricing and availability is not currently available.