MilliporeSigma
  • A DREPP protein interacted with PeaT1 from Alternaria tenuissima and is involved in elicitor-induced disease resistance in Nicotiana plants.
A DREPP protein interacted with PeaT1 from Alternaria tenuissima and is involved in elicitor-induced disease resistance in Nicotiana plants.
Journal of plant research (2018-05-08)
Fanlu Meng, Yao Xiao, Lihua Guo, Hongmei Zeng, Xiufen Yang, Dewen Qiu
ABSTRACT
PeaT1 is a proteinaceous elicitor from fungal pathogen Alternaria tenuissima. Our previous research revealed that this elicitor could induce defense response and enhance disease resistance in various plants including Nicotiana plants. However, immune activation mechanisms whereby PeaT1 elicits defense response remain unclear. In this study, the association between elicitor protein PeaT1 and the plasma membrane was assessed using the FITC (Fluorescein isothiocyanate) labeling method. A PeaT1-interacting protein was isolated via 125I-PeaT1 cross-linking and Far Western blot analyses, and designated PtBP1 (PeaT1 Binding Protein 1). From the data of Mass spectrometry (MS) and bioinformatics analysis, the 22 kDa plasma membrane protein PtBP1 was inferred to be a member of DREPP (developmentally regulated plasma membrane polypeptide) family that is induced in plants under stress conditions and might get involved in downstream signaling. For further verification of this association, Far Western blot, co-immunoprecipitation and bimolecular fluorescence complementation (BiFC) analyses were performed, showing PtBP1 could bind with PeaT1 in vitro and in vivo. Virus-induced gene silencing (VIGS) analysis exhibited that PtBP1 silencing in Nicotiana benthamiana attenuated tobacco mosaic virus (TMV) resistance compared to the tobacco rattle virus (TRV) control after PeaT1 treatment.