MilliporeSigma
  • Improved Outcomes of Cardiopulmonary Resuscitation in Rats Treated With Vagus Nerve Stimulation and Its Potential Mechanism.
Improved Outcomes of Cardiopulmonary Resuscitation in Rats Treated With Vagus Nerve Stimulation and Its Potential Mechanism.
Shock (Augusta, Ga.) (2017-08-12)
Peng Sun, Jiangang Wang, Shen Zhao, Zhengfei Yang, Ziren Tang, Navami Ravindra, Jennifer Bradley, Joseph P Ornato, Mary Ann Peberdy, Wanchun Tang
ABSTRACT
Studies have demonstrated that vagus nerve stimulation (VNS) reduces ischemia/reperfusion injury. In this study, we investigated the protective effects of VNS in a rat model of cardiopulmonary resuscitation (CPR). We further investigated whether the beneficial effects of VNS were dependent on the alpha 7 nicotinic acetylcholine receptor (α7nAChR). Forty animals were randomized into four groups and all underwent CPR (n = 10 each): CPR alone (control); VNS during CPR; α7nAChR antagonist methyllycaconitine citrate (MLA) with VNS; α7nAChR agonist 3-(2, 4-dimethoxybenzylidene) anabaseine (GTS-21 dihydrochloride) without VNS. The right vagus nerve was exteriorized in all animals. Ventricular fibrillation was induced and untreated for 8 min. Defibrillation was attempted after 8 min of CPR. VNS was initiated at the beginning of precordial chest compressions and continued for 4 h after return of spontaneous circulation (ROSC) in both the VNS and MLA groups. Hemodynamic measurements and myocardial function, including ejection fraction and myocardial performance index, were assessed at baseline, 1 and 4 h after ROSC. The neurological deficit score was measured at 24-h intervals for a total of 72 h. The heart rate was reduced in the VNS and MLA groups, while no difference was found in mean arterial pressure between the four groups. Better post-resuscitation myocardial and cerebral function and longer duration of survival were observed in the VNS-treated animals. The protective effects of VNS could be abolished by MLA and imitated by GTS-21. In addition, VNS decreased the number of electrical shocks and the duration of CPR required. VNS improves multiple outcomes after CPR.
Sigma-Aldrich
GTS-21, ≥97% (HPLC)