MilliporeSigma
  • LGR5 regulates gastric adenocarcinoma cell proliferation and invasion via activating Wnt signaling pathway.
LGR5 regulates gastric adenocarcinoma cell proliferation and invasion via activating Wnt signaling pathway.
Oncogenesis (2018-08-10)
Xiangfei Wang, Xiumin Wang, Yang Liu, Yating Dong, Yanan Wang, Muzaffer Ahmad Kassab, Wufang Fan, Xiaochun Yu, Chen Wu
ABSTRACT
LGR5 plays a critical role in tissue development and the maintenance of adult stem cells in gastrointestinal tract. However, the oncogenic role of LGR5 in the development of gastric adenocarcinoma remains elusive. Here, we show that LGR5 promotes gastric adenocarcinoma cell proliferation and metastasis. We find that knock down of LGR5 or suppression of Wnt signaling pathway by inhibitor C59 arrests gastric adenocarcinoma cell proliferation and invasion. Moreover, treatment of Wnt3a, the activator of Wnt signaling pathway, partially recovers the proliferation defect observed in LGR5 knockdown gastric adenocarcinoma cells. Moreover, LGR5 facilitates β-catenin nuclear accumulation, a surrogate marker of the activation of Wnt signaling pathway. In addition, C59 treatment suppresses transcription of Axin2 and TCF1, both of which are the target genes of β-catenin in gastric adenocarcinoma cells. Gastric adenocarcinoma cells with overexpressed LGR5 form a large quantity of visible actin filaments and pseudopods, suggesting that LGR5 significantly enhances the ability of cell movement, which might capacitate gastric adenocarcinoma cells with enhanced LGR5 expression to gain invasive and migratory properties. Taken together, our results show that LGR5 contributes to cell proliferation and invasion through the activation of Wnt/β-catenin-signaling pathway in gastric adenocarcinoma cells.
Sigma-Aldrich
MISSION® esiRNA, targeting human LGR5