MilliporeSigma
  • Giant reversible elongation upon cooling and contraction upon heating for a crosslinked cis poly(1,4-butadiene) system at temperatures below zero Celsius.
Giant reversible elongation upon cooling and contraction upon heating for a crosslinked cis poly(1,4-butadiene) system at temperatures below zero Celsius.
Scientific reports (2018-09-27)
Lu Lu, Jinbao Cao, Guoqiang Li
ABSTRACT
Polymers with reversible elongation upon cooling (EUC) and contraction upon heating (CUH) enabled applications in actuators, fasteners, dampers, grippers, swimmers, sealants, etc. With the current working temperature being limited to mainly above zero Celsius, applications for subzero Celsius environments are obstructed. In addition, current reversible actuation needs a constant tensile load, or for the best case, under zero tensile load. Reversible EUC and CUH under compressive load is almost impossible and has not been explored. In this work, a cis poly(1,4-butadiene) based system has been developed. Actuated below zero Celsius, 69% EUC occurred under a tensile load; and 6.2% EUC and 17.9% CUH occurred under 0.05 MPa compressive load. The reversible actuation was driven by both entropy and enthalpy, which was validated by a series of characterization tools.
Sigma-Aldrich
Dicumyl peroxide, 98%
Pricing and availability is not currently available.