MilliporeSigma
  • Total synthesis of (+)-batzelladine A and (-)-batzelladine D via [4 + 2]-annulation of vinyl carbodiimides with N-alkyl imines.
Total synthesis of (+)-batzelladine A and (-)-batzelladine D via [4 + 2]-annulation of vinyl carbodiimides with N-alkyl imines.
Journal of the American Chemical Society (2006-10-05)
Michael A Arnold, Kenneth A Day, Sergio G Durón, David Y Gin
ABSTRACT
A diastereoselective [4 + 2]-annulation of vinyl carbodiimides with chiral N-alkyl imines has been developed to access the stereochemically rich polycyclic guanidine cores of the batzelladine alkaloids. Application of this strategy, together with additional key steps such as long-range directed hydrogenation and diastereoselective intramolecular iodo-amination, led to highly convergent total syntheses of (-)-batzelladine D and (+)-batzelladine A with excellent stereocontrol.
Sigma-Aldrich
4-Methoxybenzyl isocyanate, 98%