MilliporeSigma
  • The IkappaB-NF-kappaB signaling module: temporal control and selective gene activation.
The IkappaB-NF-kappaB signaling module: temporal control and selective gene activation.
Science (New York, N.Y.) (2002-11-09)
Alexander Hoffmann, Andre Levchenko, Martin L Scott, David Baltimore
ABSTRACT
Nuclear localization of the transcriptional activator NF-kappaB (nuclear factor kappaB) is controlled in mammalian cells by three isoforms of NF-kappaB inhibitor protein: IkappaBalpha, -beta, and - epsilon. Based on simplifying reductions of the IkappaB-NF-kappaB signaling module in knockout cell lines, we present a computational model that describes the temporal control of NF-kappaB activation by the coordinated degradation and synthesis of IkappaB proteins. The model demonstrates that IkappaBalpha is responsible for strong negative feedback that allows for a fast turn-off of the NF-kappaB response, whereas IkappaBbeta and - epsilon function to reduce the system's oscillatory potential and stabilize NF-kappaB responses during longer stimulations. Bimodal signal-processing characteristics with respect to stimulus duration are revealed by the model and are shown to generate specificity in gene expression.
Sigma-Aldrich
I?Bβ , GST tagged human, recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution