MilliporeSigma
  • Clinical impact of oral antidiabetic medications in heart failure patients.
Clinical impact of oral antidiabetic medications in heart failure patients.
Heart failure reviews (2018-01-24)
Alberto Palazzuoli, Elena Ceccarelli, Gaetano Ruocco, Ranuccio Nuti
ABSTRACT
Heart failure (HF) is a common complication in patients with type 2 diabetes and it is closely associated with high morbidity and mortality rate. The incidence of cardiovascular events in patients with diabetes is related to high levels of glycemia, expressed by increase of HbA1c levels. However, there is little evidence to indicate that glycemic control can reduce the incidence of HF events in this population. Recently, several new antidiabetic drugs have been proposed although the exact clinical impact on heart failure occurrence and deterioration is under debate. Most common oral antidiabetic medication such as SGLT2, GLP-1 receptor agonist, metformin, and DPP4 inhibitor revealed peculiar metabolic and biomolecular signal effects. Moreover, the negative effects of thiazolidinediones on HF prognosis, on cardiac function, and exercise tolerance is of great interest. Conversely, several studies on GLP-1RA have highlighted many positive effects on cardiac myocytes, reducing apoptosis through cAMP/PKA/CRCB-mediated pathways protecting against oxidative stress. DPP-4 inhibitors have a controversial effect: saxagliptin and alogliptin may increase the risk of HF as opposed to vildagliptin and sitagliptin. Metformin increases myocardial ATP levels due to activation of 5-AMPK and this could explain the positive link between the drug and events rate reduction in diabetic patients with HF. The more interesting class of new drugs is SGLT2 inhibitors, that seems to have a positive effect on cardiac function by 38% reduction of HF incidence and mortality with empagliflozin treatment. In this review, we would analyze the specific effects of each class so as to better elucidate the clinical impact of antidiabetic drug on HF for guiding the clinicians in the choice of a best individualized therapy.
Sigma-Aldrich
Vildagliptin, ≥98% (HPLC)
Pricing and availability is not currently available.