MilliporeSigma
  • A nucleosomal function for IkappaB kinase-alpha in NF-kappaB-dependent gene expression.
A nucleosomal function for IkappaB kinase-alpha in NF-kappaB-dependent gene expression.
Nature (2003-06-06)
Vasiliki Anest, Julie L Hanson, Patricia C Cogswell, Kris A Steinbrecher, Brian D Strahl, Albert S Baldwin
ABSTRACT
NF-kappaB is a principal transcriptional regulator of diverse cytokine-mediated processes and is tightly controlled by the IkappaB kinase complex (IKK-alpha/beta/gamma). IKK-beta and IKK-gamma are critical for cytokine-induced NF-kappaB function, whereas IKK-alpha is thought to be involved in other regulatory pathways. However, recent data suggest a role for IKK-alpha in NF-kappaB-dependent gene expression in response to cytokine treatment. Here we demonstrate nuclear accumulation of IKK-alpha after cytokine exposure, suggesting a nuclear function for this protein. Consistent with this, chromatin immunoprecipitation (ChIP) assays reveal that IKK-alpha was recruited to the promoter regions of NF-kappaB-regulated genes on stimulation with tumour-necrosis factor-alpha. Notably, NF-kappaB-regulated gene expression is suppressed by the loss of IKK-alpha and this correlates with a complete loss of gene-specific phosphorylation of histone H3 on serine 10, a modification previously associated with positive gene expression. Furthermore, we show that IKK-alpha can directly phosphorylate histone H3 in vitro, suggesting a new substrate for this kinase. We propose that IKK-alpha is an essential regulator of NF-kappaB-dependent gene expression through control of promoter-associated histone phosphorylation after cytokine exposure. These findings provide additional insight into the role of the IKK complex in NF-kappaB-regulated gene expression.
Sigma-Aldrich
IKK α , active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Pricing and availability is not currently available.