MilliporeSigma
BRCA1 physically and functionally interacts with ATF1.
The Journal of biological chemistry (2000-08-18)
Y Houvras, M Benezra, H Zhang, J J Manfredi, B L Weber, J D Licht
ABSTRACT
BRCA1, a breast and ovarian cancer susceptibility gene, encodes a 220-kDa protein whose precise biochemical function remains unclear. BRCA1 contains an N-terminal RING finger that mediates protein-protein interaction. The C-terminal domain of BRCA1 (BRCT) can activate transcription and interacts with RNA polymerase holoenzyme. Using the yeast two-hybrid system, we identified an interaction between the BRCA1 RING finger and ATF1, a member of the cAMP response element-binding protein/activating transcription factor (CREB/ATF) family. We demonstrate that BRCA1 and ATF1 can physically associate in vitro, in yeast, and in human cells. BRCA1 stimulated transcription from a cAMP response element reporter gene in transient transfections. BRCA1 also stimulated transcription from a natural promoter, that of tumor necrosis factor-alpha, in a manner dependent on the integrity of the cAMP response element. These results implicate BRCA1 in transcriptional activation of ATF1 target genes, some of which are involved in the transcriptional response to DNA damage.