MilliporeSigma
  • Involvement of tyrosine phosphorylation in the positive inotropic effect produced by H(1)-receptors with histamine in guinea-pig left atrium.
Involvement of tyrosine phosphorylation in the positive inotropic effect produced by H(1)-receptors with histamine in guinea-pig left atrium.
British journal of pharmacology (2000-06-24)
Y Akaishi, Y Hattori, K Yoshimoto, A Kitabatake, K Yasuda, M Kanno
ABSTRACT
We investigated the effect of stimulation of H(1)-receptors with histamine on protein tyrosine phosphorylation levels in guinea-pig left atrium and evaluated the influences of tyrosine kinase inhibitors on the positive inotropic effect mediated by H(1)-receptors in this tissue. Histamine induced an increase in tyrosine phosphorylation in four main clusters of proteins with apparent molecular weights of 25, 35, 65 and 150 kDa. Tyrosine phosphorylation of these proteins attained a peak around 2 - 3 min following histamine stimulation and then declined to or below basal levels. Histamine-induced protein tyrosine phosphorylation was antagonized by the H(1)-receptor antagonists mepyramine (1 microM) and chlorpheniramine (1 microM), but not by the H(2)-receptor antagonist cimetidine (10 microM). The positive inotropic effect of histamine was depressed in a concentration-dependent manner by the tyrosine kinase inhibitors tyrphostin A25 (50 to 100 microM) and genistein (10 to 50 microM) but not by the inactive genistein analogue daidzein (50 microM). The positive inotropic effect of isoprenaline was unchanged by tyrphostin A25 and genistein. At a concentration of 1 microM histamine produced a dual-component positive inotropic response composed of an initial increasing phase and a second and late developing, greater positive inotropic phase. Treatment with tyrphostin A25 (100 microM) and genistein (50 microM), but not daidzein (50 microM), significantly attenuated the two components of the inotropic response, although genistein suppressed the initial component more markedly than the late component. We conclude that increased protein tyrosine phosphorylation may play an important role in initiating at least some part of the positive inotropic effect of H(1)-receptor stimulation in guinea-pig left atrium.
Sigma-Aldrich
S-(+)-Chlorpheniramine maleate salt
Pricing and availability is not currently available.