MilliporeSigma
  • Fabrication of single-chain nanoparticles through the dimerization of pendant anthracene groups via photochemical upconversion.
Fabrication of single-chain nanoparticles through the dimerization of pendant anthracene groups via photochemical upconversion.
Dalton transactions (Cambridge, England : 2003) (2018-06-23)
Tamuka Chidanguro, Danielle R Blank, Alexandra Garrett, Cassandra M Reese, Jacob M Schekman, Xinjun Yu, Derek L Patton, Neil Ayres, Yoan C Simon
ABSTRACT
We report on the use of visible light as the driving force for the intramolecular dimerization of pendant anthracene groups on a methacrylic polymer to induce the formation of single-chain nanoparticles (SCNPs). Using a 532 nm green laser light source and platinum octaethylporphyrin as a sensitizer, we first demonstrated the use of TTA-UC to dimerize monomeric anthracene, and subsequently applied this concept to dilute poly((methyl methacrylate)-stat-(anthracenyl methacrylate)) samples. A combination of triple-detection size-exclusion chromatography, atomic force microscopy, and UV-visible spectroscopy confirmed the formation of the SCNPs. This report pioneers the use of TTA-UC to drive photochemical reactions in polymeric systems, and showcases the potential for TTA-UC in the development of nanoobjects.
Sigma-Aldrich
9-Anthracenemethanol, 97%
Pricing and availability is not currently available.