MilliporeSigma
  • LAT1-Targeting Thermoresponsive Fluorescent Polymer Probes for Cancer Cell Imaging.
LAT1-Targeting Thermoresponsive Fluorescent Polymer Probes for Cancer Cell Imaging.
International journal of molecular sciences (2018-06-06)
Minami Matsuura, Mariko Ohshima, Yuki Hiruta, Tomohiro Nishimura, Kenichi Nagase, Hideko Kanazawa
ABSTRACT
L-type amino acid transporter 1 (LAT1) is more highly expressed in cancer cells compared with normal cells. LAT1 targeting probes would therefore be a promising tool for cancer cell imaging. In this study, LAT1-targeting thermoresponsive fluorescent polymer probes based on poly(N-isopropylacrylamide-co-N,N-dimethylacrylamide) (P(NIPAAm-co-DMAAm)) were synthesized and their affinity for LAT1 was evaluated. The synthesized polymer probes interacted with LAT1 on HeLa cells, and inhibition of l-[³H]-leucine, one of the substrates for LAT1 uptake, was investigated. l-Tyrosine-conjugated P(NIPAAm-co-DMAAm) inhibited the uptake of l-[³H]-leucine, while P(NIPAAm-co-DMAAm) and l-phenylalanine-conjugated P(NIPAAm-co-DMAAm) did not. This result indicated that l-tyrosine-conjugated polymer has a high affinity for LAT1. The fluorescent polymer probes were prepared by modification of a terminal polymer group with fluorescein-5-maleimide (FL). Above the polymer transition temperature, cellular uptake of the polymer probes was observed because the polymers became hydrophobic, which enhanced the interaction with the cell membrane. Furthermore, quantitative analysis of the fluorescent probe using flow cytometry indicated that l-tyrosine-conjugated P(NIPAAm-co-DMAAm)-FL shows higher fluorescence intensity earlier than P(NIPAAm-co-DMAAm)-FL. The result suggested that cellular uptake was promoted by the LAT1 affinity site. The developed LAT1-targeting thermoresponsive fluorescent polymer probes are expected to be useful for cancer cell imaging.
Sigma-Aldrich
N,N-Dimethylacrylamide, 99%, contains 500 ppm monomethyl ether hydroquinone as inhibitor
Pricing and availability is not currently available.
Sigma-Aldrich
4-Cyano-4-(phenylcarbonothioylthio)pentanoic acid
Pricing and availability is not currently available.