MilliporeSigma
Dynamics of human protein kinase Aurora A linked to drug selectivity.
eLife (2018-06-15)
Warintra Pitsawong, Vanessa Buosi, Renee Otten, Roman V Agafonov, Adelajda Zorba, Nadja Kern, Steffen Kutter, Gunther Kern, Ricardo Ap Pádua, Xavier Meniche, Dorothee Kern
ABSTRACT
Protein kinases are major drug targets, but the development of highly-selective inhibitors has been challenging due to the similarity of their active sites. The observation of distinct structural states of the fully-conserved Asp-Phe-Gly (DFG) loop has put the concept of conformational selection for the DFG-state at the center of kinase drug discovery. Recently, it was shown that Gleevec selectivity for the Tyr-kinase Abl was instead rooted in conformational changes after drug binding. Here, we investigate whether protein dynamics after binding is a more general paradigm for drug selectivity by characterizing the binding of several approved drugs to the Ser/Thr-kinase Aurora A. Using a combination of biophysical techniques, we propose a universal drug-binding mechanism, that rationalizes selectivity, affinity and long on-target residence time for kinase inhibitors. These new concepts, where protein dynamics in the drug-bound state plays the crucial role, can be applied to inhibitor design of targets outside the kinome.
Sigma-Aldrich
5-Fluoro-DL-tryptophan, powder or crystals
Pricing and availability is not currently available.
Sigma-Aldrich
5-Fluoro-L-tryptophan, ≥98.0% (HPLC)
Pricing and availability is not currently available.