MilliporeSigma
  • Light-Triggered Radiochemical Synthesis: A Novel 18F-Labelling Strategy Using Photoinducible Click Reaction to Prepare PET Imaging Probes.
Light-Triggered Radiochemical Synthesis: A Novel 18F-Labelling Strategy Using Photoinducible Click Reaction to Prepare PET Imaging Probes.
Contrast media & molecular imaging (2018-07-27)
Ji Hae Choi, Doori Oh, In Sun Kim, Hyeon-Soo Kim, Minjoo Kim, Eun-Mi Kim, Seok Tae Lim, Myung-Hee Sohn, Dong Hyun Kim, Hwan-Jeong Jeong
ABSTRACT
Novel probe development for positron emission tomography (PET) is leading to expanding the scope of molecular imaging. To begin responding to challenges, several biomaterials such as natural products and small molecules, peptides, engineered proteins including affibodies, and antibodies have been used in the development of targeted molecular imaging probes. To prepare radiotracers, a few bioactive materials are unique challenges to radiolabelling because of their complex structure, poor stability, poor solubility in aqueous or chemical organic solutions, and sensitivity to temperature and nonphysiological pH. To overcome these challenges, we developed a new radiolabelling strategy based on photoactivated 1,3-dipolar cycloaddition between alkene dipolarophile and tetrazole moiety containing compounds. Herein, we describe a light-triggered radiochemical synthesis via photoactivated click reaction to prepare 18F-radiolabelled PET tracers using small molecular and RGD peptide.
Sigma-Aldrich
Benzenesulfonyl hydrazide, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Allyloxyethanol, 98%
Pricing and availability is not currently available.