MilliporeSigma
  • Dual modulation on glial cells by tetrahydroxystilbene glucoside protects against dopamine neuronal loss.
Dual modulation on glial cells by tetrahydroxystilbene glucoside protects against dopamine neuronal loss.
Journal of neuroinflammation (2018-05-29)
Yanzhen Zhou, Guoqing Wang, Daidi Li, Yanying Wang, Qin Wu, Jingshan Shi, Feng Zhang
ABSTRACT
Microglia-mediated neuroinflammation is recognized to mainly contribute to the pathogenesis of Parkinson's disease (PD). Tetrahydroxystilbene glucoside (TSG) has been proved to be beneficial for health with a great number of pharmacological properties. We examined the effects of TSG against dopamine (DA) neuronal loss towards development of a PD treatment strategy. Substantia nigral stereotaxic single injection of lipopolysaccharide (LPS)-induced rat DA neuronal damage was employed to investigate TSG-produced neuroprotection. In addition, primary rat midbrain neuron-glia co-cultures were performed to explore the underlying mechanisms. Daily intraperitoneal injection of TSG for seven consecutive days significantly attenuated LPS-induced loss of DA neurons in the substantia nigra. In addition, glia-dependent mechanisms were responsible for TSG-mediated neuroprotection. First, TSG ameliorated microglia-mediated neuroinflammation and the subsequent production of various pro-inflammatory and neurotoxic factors. Second, astroglial neurotrophic factor neutralization weakened TSG-mediated neuroprotection, showing that TSG was protective in part via increasing astroglia-derived neurotrophic factor secretion. TSG protects DA neurons against LPS-induced neurotoxicity through dual modulation on glial cells by attenuating microglia-mediated neuroinflammation and enhancing astroglia-derived neurotrophic effects. These findings might open new alternative avenues for PD treatment.
Sigma-Aldrich
Octoclothepin maleate salt, solid
Sigma-Aldrich
Leu-Leu