MilliporeSigma
  • Amaryllidaceae Alkaloids as Potential Glycogen Synthase Kinase-3β Inhibitors.
Amaryllidaceae Alkaloids as Potential Glycogen Synthase Kinase-3β Inhibitors.
Molecules (Basel, Switzerland) (2018-03-22)
Daniela Hulcová, Kateřina Breiterová, Tomáš Siatka, Kamila Klímová, Lara Davani, Marcela Šafratová, Anna Hošťálková, Angela De Simone, Vincenza Andrisano, Lucie Cahlíková
ABSTRACT
Glycogen synthase kinase-3β (GSK-3β) is a multifunctional serine/threonine protein kinase that was originally identified as an enzyme involved in the control of glycogen metabolism. It plays a key role in diverse physiological processes including metabolism, the cell cycle, and gene expression by regulating a wide variety of well-known substances like glycogen synthase, tau-protein, and β-catenin. Recent studies have identified GSK-3β as a potential therapeutic target in Alzheimer´s disease, bipolar disorder, stroke, more than 15 types of cancer, and diabetes. GSK-3β is one of the most attractive targets for medicinal chemists in the discovery, design, and synthesis of new selective potent inhibitors. In the current study, twenty-eight Amaryllidaceae alkaloids of various structural types were studied for their potency to inhibit GSK-3β. Promising results have been demonstrated by alkaloids of the homolycorine-{9-O-demethylhomolycorine (IC50 = 30.00 ± 0.71 µM), masonine (IC50 = 27.81 ± 0.01 μM)}, and lycorine-types {caranine (IC50 = 30.75 ± 0.04 μM)}.
Sigma-Aldrich
SB 415286, ≥98% (HPLC)
Pricing and availability is not currently available.