MilliporeSigma
  • Gomisin A modulates aging progress via mitochondrial biogenesis in human diploid fibroblast cells.
Gomisin A modulates aging progress via mitochondrial biogenesis in human diploid fibroblast cells.
Clinical and experimental pharmacology & physiology (2018-01-11)
Jeong-Seok Kim, Seon-Hwa Jeong, Sin-Hee Han, Ho-Keun Yi
ABSTRACT
Gomisin A from the fruit of Schisandra chinensis has many pharmacological properties, including hepato-protective, anti-diabetic, and anti-oxidative stress. However, the potential benefit of gomisin A is still not well understood, especially in aging progression. Therefore, the aim of this study was to clarify whether the promotion of mitochondrial biogenesis and autophagy of gomisin A affects anti-aging progression, and its mechanism. Intermediate (PD32) human diploid fibroblast (HDF) cells were brought to stress-induced premature senescence (SIPS) using hydrogen peroxide. Gomisin A inhibited reactive oxygen species production even in the SIPS-HDF cells. Gomisin A was also able to attenuate the activity of senescence-associated β-galactosidase and the production of pro-inflammatory molecules in the SIPS as well as aged HDF cells. The antioxidant activity of gomisin A was determined by recovering the Cu/Zn, Mn-SOD, and HO-1 expression in the SIPS-HDF cells. In mechanistic aspect, gomisin A inhibited the mitogen-activated protein kinase pathway and the translocation of nuclear factor kappa B to the nucleus. In addition, gomisin A promoted the autophagy and mitochondrial biogenesis factors through the translocation of nuclear factor erythroid 2-related factor-2, and inhibited aging progression in the SIPS-HDF cells. In summary, the enhanced properties of mitochondrial biogenesis and autophagy of gomisin A has a benefit to control age-related molecules against SIPS-induced chronic oxidative stress, and gomisin A may be a potential therapeutic compound for the enhancement of intracellular homeostasis to aging progression.
Sigma-Aldrich
Gomisin A
Pricing and availability is not currently available.